siRNA Details
Gene Symbol: OR7A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017506.1 | NP_059976.1 | 1 | 407 | ||
XM_011527912.2 | XP_011526214.1 | 2 | 432 | |||
XM_011527913.2 | XP_011526215.1 | 2 | 358 | |||
XM_017026583.1 | XP_016882072.1 | 2 | 321 | |||
GenBank mRNA | AK312475.1 | - | 2 | 273 | ||
BC104809.1 | - | 1 | 242 | 8143812 |
Gene Symbol: DAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HQ179936.1 | - | 4 | 341 | ||
HQ179937.1 | - | 5 | 433 |
Gene Symbol: LOC650293 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040071.1 | NP_001035160.1 | 1 | 85 |
Gene Symbol: MGC72080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002822.3 | 4 | 454 | |||
GenBank mRNA | BC029615.1 | - | 4 | 464 | ||
BC042035.1 | - | 3 | 393 |
Gene Symbol: OR7A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005190.1 | NP_001005190.1 | 1 | 184 |
Gene Symbol: OR7A17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030901.1 | NP_112163.1 | 1 | 184 | ||
GenBank mRNA | BC101587.1 | - | 1 | 281 | 8069093 | |
BC117357.1 | - | 1 | 281 | 40125908 |
Gene Symbol: OR7C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198944.1 | NP_945182.1 | 1 | 184 | ||
XM_005259853.3 | XP_005259910.1 | 3 | 688 | |||
XM_011527915.2 | XP_011526217.1 | 5 | 1109 | |||
GenBank mRNA | BC117212.1 | - | 1 | 259 | 40125763 |
Gene Symbol: OR7C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012377.1 | NP_036509.1 | 1 | 184 |
Gene Symbol: OR7D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175883.2 | NP_787079.1 | 1 | 372 | ||
GenBank mRNA | AK095468.1 | - | 1 | 372 |
Gene Symbol: OR7D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005191.2 | NP_001005191.1 | 1 | 218 | ||
GenBank mRNA | BC137147.1 | - | 1 | 218 | 9021144 | |
BC137162.1 | - | 1 | 218 | 9021159 |
Gene Symbol: OR7E14P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045002.1 | 1 | 353 | |||
GenBank mRNA | BC008657.2 | - | 1 | 208 |
Gene Symbol: OR7E154P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK122996.1 | - | 1 | 3756 |
Gene Symbol: OR7E24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079935.1 | NP_001073404.1 | 1 | 238 | ||
GenBank mRNA | BC146920.1 | - | 1 | 352 | 9056848 |
Gene Symbol: OR7E2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045004.1 | 1 | 124 |
Gene Symbol: OR7E37P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002163.3 | 1 | 343 |
Gene Symbol: OR7E47P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120437.1 | 3 | 697 | |||
NR_120439.1 | 2 | 220 | ||||
NR_120440.1 | 2 | 197 | ||||
GenBank mRNA | AA229829.1 | - | 2 | 196 | ||
BC042060.1 | - | 3 | 697 |
Gene Symbol: OR7E5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027688.2 | 4 | 640 | |||
GenBank mRNA | AK055955.1 | - | 4 | 640 |
Gene Symbol: OR7G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005193.1 | NP_001005193.1 | 1 | 247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
