siRNA Details
Gene Symbol: MAN1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016219.4 | NP_057303.2 | 11 | 1743 | ||
NR_045720.1 | 11 | 1718 | ||||
NR_045721.1 | 12 | 1874 | ||||
XM_006716945.3 | XP_006717008.1 | 11 | 1808 | |||
XM_017014239.1 | XP_016869728.1 | 12 | 1720 | |||
GenBank mRNA | AB209276.1 | - | 2 | 2463 | ||
AF145732.1 | - | 11 | 1660 | |||
AF148509.1 | - | 11 | 1720 | |||
AK074559.1 | - | 3 | 307 | |||
AK074699.1 | - | 9 | 1318 | |||
AK094878.1 | - | 3 | 1749 | |||
AK126926.1 | - | 4 | 3777 | |||
AK127005.1 | - | 4 | 2842 | |||
AK128033.1 | - | 3 | 4028 | |||
AK128257.1 | - | 4 | 2920 | |||
AK128341.1 | - | 4 | 3590 | |||
AK130143.1 | - | 6 | 887 | |||
AK298480.1 | - | 12 | 1678 | |||
AK299043.1 | - | 11 | 1590 | |||
AK315797.1 | - | 11 | 1660 | |||
AL110221.1 | - | 4 | 465 | |||
AY358465.1 | - | 11 | 1692 | |||
BC002953.1 | - | 11 | 1676 | 3533651 | ||
BC006079.1 | - | 11 | 1685 | 3959196 | ||
CR749534.1 | - | 17 | 6226 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
