siRNA Details
Gene Symbol: MAN1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016219.4 | NP_057303.2 | 13 | 2127 | ||
NR_045720.1 | 13 | 2102 | ||||
NR_045721.1 | 14 | 2258 | ||||
XM_006716945.3 | XP_006717008.1 | 12 | 2917 | |||
XM_017014239.1 | XP_016869728.1 | 13 | 2829 | |||
GenBank mRNA | AK128033.1 | - | 5 | 4412 | ||
AK128257.1 | - | 6 | 3304 | |||
AK128341.1 | - | 6 | 3974 | |||
AK298480.1 | - | 14 | 2062 | |||
BC002953.1 | - | 13 | 2060 | 3533651 | ||
CR749534.1 | - | 19 | 6610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
