siRNA Details
Gene Symbol: MAN1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016219.4 | NP_057303.2 | 13 | 2752 | ||
NR_045720.1 | 13 | 2727 | ||||
NR_045721.1 | 14 | 2883 | ||||
XM_006716945.3 | XP_006717008.1 | 12 | 3542 | |||
XM_017014239.1 | XP_016869728.1 | 13 | 3454 | |||
GenBank mRNA | AB209276.1 | - | 4 | 3472 | ||
AF145732.1 | - | 13 | 2669 | |||
AF148509.1 | - | 13 | 2728 | |||
AK074559.1 | - | 5 | 1316 | |||
AK074699.1 | - | 11 | 2327 | |||
AK094878.1 | - | 5 | 2758 | |||
AK126926.1 | - | 6 | 4786 | |||
AK127005.1 | - | 6 | 3851 | |||
AK128033.1 | - | 5 | 5037 | |||
AK128257.1 | - | 6 | 3929 | |||
AK128341.1 | - | 7 | 4513 | |||
AK130143.1 | - | 8 | 1897 | |||
AL110221.1 | - | 5 | 2199 | |||
AY358465.1 | - | 13 | 2701 | |||
BC002953.1 | - | 13 | 2685 | 3533651 | ||
BC006079.1 | - | 13 | 2694 | 3959196 | ||
CR749534.1 | - | 19 | 7235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
