siRNA Details
Gene Symbol: HBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000558.4 | NP_000549.1 | 3 | 578 | ||
GenBank mRNA | AI064720.1 | - | 2 | 336 | ||
AK223392.1 | - | 3 | 555 | |||
BC005931.1 | - | 3 | 549 | 4047053 | ||
BC032122.2 | - | 3 | 535 | 4849981 | ||
BC050661.1 | - | 3 | 535 | 5209215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
