siRNA Details
Gene Symbol: ATP6AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001183.5 | NP_001174.2 | 2 | 278 | ||
XM_011531179.1 | XP_011529481.1 | 2 | 241 | |||
GenBank mRNA | AK090462.1 | - | 2 | 212 | ||
AK091083.1 | - | 1 | 259 | |||
AK130616.1 | - | 2 | 233 | |||
AK289452.1 | - | 2 | 272 | |||
AL136851.1 | - | 2 | 212 | |||
BC000724.2 | - | 2 | 235 | 3506925 | ||
BX461330.2 | - | 2 | 247 | |||
DA921677.1 | - | 2 | 278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
