siRNA Details
Gene Symbol: ATP6AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001183.5 | NP_001174.2 | 4 | 486 | ||
XM_011531179.1 | XP_011529481.1 | 5 | 816 | |||
GenBank mRNA | AK026519.1 | - | 1 | 44 | ||
AK075284.1 | - | 4 | 450 | |||
AK090462.1 | - | 4 | 2903 | |||
AK091083.1 | - | 3 | 467 | |||
AK130616.1 | - | 4 | 441 | |||
AK289452.1 | - | 4 | 480 | |||
AL136851.1 | - | 4 | 420 | |||
BC000724.2 | - | 4 | 443 | 3506925 | ||
BX461330.2 | - | 4 | 455 | |||
D16469.1 | - | 3 | 1211 | |||
DA921677.1 | - | 4 | 486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
