siRNA Details
Gene Symbol: FOXP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148900.3 | NP_683698.2 | 5 | 790 | ||
NR_033766.1 | 8 | 952 | ||||
GenBank mRNA | AF454830.1 | - | 10 | 1202 | ||
AK131266.1 | - | 8 | 936 | |||
AK296957.1 | - | 5 | 670 | |||
CR749236.1 | - | 10 | 968 | |||
KJ535079.1 | - | 4 | 416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
