siRNA Details
Gene Symbol: FOXP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172766.2 | NP_001166237.1 | 4, 5 | 778 | ||
NM_001172767.2 | NP_001166238.1 | 4, 5 | 489 | |||
NM_014491.3 | NP_055306.1 | 4, 5 | 778 | |||
NM_148898.3 | NP_683696.2 | 5, 6 | 853 | |||
NM_148899.3 | NP_683697.2 | 3, 4 | 414 | |||
NR_033767.1 | 5, 6 | 836 | ||||
XM_017012801.1 | XP_016868290.1 | 7, 8 | 1459 | |||
GenBank mRNA | AF337817.1 | - | 4, 5 | 554 | ||
AF454830.1 | - | 11 | 1259 | |||
AF467252.1 | - | 4, 5 | 505 | |||
AF467253.1 | - | 3, 4 | 416 | |||
AF467257.1 | - | 4, 5 | 456 | |||
AF467258.1 | - | 3, 4 | 401 | |||
AF467259.1 | - | 4, 5 | 476 | |||
AY144615.1 | - | 5, 6 | 848 | |||
BC126104.1 | - | 5, 6 | 510 | 8991820 | ||
BC143866.1 | - | 5, 6 | 510 | 9052382 | ||
BC143867.1 | - | 4, 5 | 452 | 9052383 | ||
DQ778626.1 | - | 1, 2 | 128 | |||
KJ534839.1 | - | 4, 5 | 479 | |||
KJ535079.1 | - | 4 | 617 | |||
KJ535086.1 | - | 3, 4 | 403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
