siRNA Details
Gene Symbol: PLGLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003506.3 | 1 | 83 | |||
GenBank mRNA | DA634686.1 | - | 1 | 83 |
Gene Symbol: LPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005577.2 | NP_005568.2 | 2 | 144 | ||
GenBank mRNA | X06290.1 | - | 69 |
Gene Symbol: PLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000301.3 | NP_000292.1 | 1 | 136 | ||
NM_001168338.1 | NP_001161810.1 | 1 | 136 | |||
GenBank mRNA | AK298338.1 | - | 1 | 85 | ||
AK308036.1 | - | 1 | 87 | |||
AK312966.1 | - | 1 | 87 | |||
BC060513.1 | - | 1 | 62 | 4609795 | ||
DR004070.1 | - | 1 | 143 | |||
M74220.1 | - | 1 | 73 | |||
X05199.1 | - | 1 | 78 |
Gene Symbol: PLGLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032392.2 | NP_001027564.1 | 1 | 87 | ||
GenBank mRNA | BC070296.1 | - | 1 | 66 | ||
BC107767.1 | - | 1 | 62 | |||
BC109248.2 | - | 1 | 63 | 40022876 |
Gene Symbol: PLGLB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002665.4 | NP_002656.1 | 1 | 91 | ||
GenBank mRNA | BC005379.1 | - | 1 | 87 | 3934860 | |
BC022294.1 | - | 1 | 93 | 4716526 | ||
M93143.2 | - | 1 | 91 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
