siRNA Details
Gene Symbol: CSNK1A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145203.5 | NP_660204.2 | 1 | 710 | ||
GenBank mRNA | BC028723.2 | - | 1 | 712 | 5266487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
