siRNA Details
Gene Symbol: NEU4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025617.1 | - | 5 | 2025 | ||
AK091038.1 | - | 5 | 2270 | |||
AK096992.1 | - | 2 | 2885 | |||
AK289421.1 | - | 4 | 1649 | |||
AK290076.1 | - | 4 | 1632 | |||
AK300969.1 | - | 4 | 1645 | |||
AY203952.1 | - | 4 | 1627 | |||
BC012899.1 | - | 4 | 2046 | 4156395 | ||
BC105974.1 | - | 4 | 1608 | 4944243 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
