siRNA Details
Gene Symbol: NEU4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167599.2 | NP_001161071.1 | 4 | 2118 | ||
NM_001167600.2 | NP_001161072.1 | 4 | 1795 | |||
NM_001167601.2 | NP_001161073.1 | 4 | 1798 | |||
NM_001167602.2 | NP_001161074.1 | 5 | 2296 | |||
NM_080741.3 | NP_542779.2 | 4 | 2115 | |||
GenBank mRNA | AK025617.1 | - | 5 | 2048 | ||
AK096992.1 | - | 2 | 2908 | |||
AK289421.1 | - | 4 | 1672 | |||
AK290076.1 | - | 4 | 1655 | |||
AK300969.1 | - | 4 | 1668 | |||
AY203952.1 | - | 4 | 1650 | |||
BC012899.1 | - | 4 | 2069 | 4156395 | ||
BC105974.1 | - | 4 | 1631 | 4944243 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
