siRNA Details
Gene Symbol: TMEM86B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173804.4 | NP_776165.3 | 1 | 218 | ||
GenBank mRNA | BC023000.1 | - | 1 | 218 | 5000011 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
