siRNA Details
Gene Symbol: FAM105A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019018.2 | NP_061891.1 | 8 | 3820 | ||
GenBank mRNA | AK125648.1 | - | 1 | 1112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
