siRNA Details
Gene Symbol: SLC35F6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017877.3 | NP_060347.2 | 6 | 1800 | ||
GenBank mRNA | BX647807.1 | - | 5 | 1718 | ||
BX648017.1 | - | 5 | 1578 |
Gene Symbol: PCTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648199.1 | - | 1 | 2077 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
