siRNA Details
Gene Symbol: GOLGA8N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282494.1 | NP_001269423.1 | 4 | 382 | ||
GenBank mRNA | AK310041.1 | - | 2 | 146 |
Gene Symbol: GOLGA8O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277308.1 | NP_001264237.1 | 4 | 382 | ||
XM_011521988.2 | XP_011520290.1 | 4 | 1067 | |||
XM_011521989.2 | XP_011520291.1 | 4 | 1071 | |||
GenBank mRNA | DB068178.1 | - | 4 | 382 |
Gene Symbol: GOLGA8R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282484.1 | NP_001269413.1 | 4 | 382 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
