siRNA Details
Gene Symbol: GOLGA8M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282468.1 | NP_001269397.1 | 7 | 525 | ||
XM_005268284.4 | XP_005268341.1 | 7 | 1324 | |||
XM_005268285.4 | XP_005268342.1 | 7 | 1324 | |||
XM_006720654.3 | XP_006720717.1 | 7 | 468 | |||
XM_011521951.2 | XP_011520253.1 | 8 | 893 | |||
XM_011521952.2 | XP_011520254.1 | 7 | 1324 | |||
XM_011521953.2 | XP_011520255.1 | 7 | 1324 | |||
XM_011521954.2 | XP_011520256.1 | 7 | 458 |
Gene Symbol: GOLGA8IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024074.1 | 6 | 726 | |||
GenBank mRNA | AK093104.1 | - | 6 | 726 |
Gene Symbol: GOLGA8J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282472.1 | NP_001269401.1 | 7 | 525 | ||
XM_006720651.3 | XP_006720714.1 | 7 | 545 | |||
XM_011521948.2 | XP_011520250.1 | 7 | 853 | |||
XM_011521950.2 | XP_011520252.1 | 7 | 851 | |||
GenBank mRNA | DB054802.1 | - | 7 | 414 | ||
DB098469.1 | - | 7 | 545 |
Gene Symbol: GOLGA8K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282493.1 | NP_001269422.1 | 7 | 525 |
Gene Symbol: GOLGA8N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282494.1 | NP_001269423.1 | 7 | 525 |
Gene Symbol: GOLGA8O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277308.1 | NP_001264237.1 | 7 | 525 | ||
XM_011521988.2 | XP_011520290.1 | 7 | 1210 | |||
XM_011521989.2 | XP_011520291.1 | 7 | 1214 | |||
GenBank mRNA | BQ477899.1 | - | 2 | 55 | ||
DB068178.1 | - | 7 | 525 |
Gene Symbol: GOLGA8R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282484.1 | NP_001269413.1 | 7 | 525 |
Gene Symbol: GOLGA8T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033933.1 | 7 | 545 | |||
GenBank mRNA | AK310526.1 | - | 7 | 545 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
