siRNA Details
Gene Symbol: RGPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078170.2 | NP_001071638.2 | 4 | 325 | ||
XM_017004845.1 | XP_016860334.1 | 8 | 776 | |||
XM_017004846.1 | XP_016860335.1 | 4 | 447 | |||
XM_017004847.1 | XP_016860336.1 | 4 | 444 | |||
XM_017004848.1 | XP_016860337.1 | 8 | 776 | |||
XM_017004849.1 | XP_016860338.1 | 8 | 776 | |||
XM_017004850.1 | XP_016860339.1 | 8 | 776 | |||
GenBank mRNA | AK302429.1 | - | 4 | 325 |
Gene Symbol: RANBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006267.4 | NP_006258.3 | 4 | 416 | ||
XM_005264002.2 | XP_005264059.1 | 4 | 451 | |||
XM_005264003.2 | XP_005264060.1 | 4 | 379 | |||
XM_005264004.2 | XP_005264061.1 | 4 | 379 | |||
XM_005264005.4 | XP_005264062.1 | 4 | 439 | |||
XM_005264007.2 | XP_005264064.1 | 4 | 379 | |||
XM_011511575.2 | XP_011509877.1 | 4 | 354 | |||
XM_011511576.2 | XP_011509878.1 | 4 | 379 | |||
XM_011511578.2 | XP_011509880.1 | 4 | 421 | |||
XM_017004623.1 | XP_016860112.1 | 4 | 379 | |||
XM_017004624.1 | XP_016860113.1 | 4 | 379 | |||
XM_017004625.1 | XP_016860114.1 | 4 | 451 | |||
GenBank mRNA | AB209483.1 | - | 1 | 32 | ||
D42063.1 | - | 4 | 417 |
Gene Symbol: RGPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024457.3 | NP_001019628.3 | 4 | 279 | ||
XM_011532845.2 | XP_011531147.1 | 5 | 478 | |||
XM_011532849.2 | XP_011531151.1 | 5 | 478 | |||
XM_011532850.1 | XP_011531152.1 | 5 | 478 | |||
XM_011532851.2 | XP_011531153.1 | 5 | 478 | |||
XM_011532852.2 | XP_011531154.1 | 5 | 478 | |||
XM_017004103.1 | XP_016859592.1 | 5 | 478 | |||
XM_017004104.1 | XP_016859593.1 | 4 | 595 | |||
XM_017004105.1 | XP_016859594.1 | 4 | 443 | |||
XM_017004106.1 | XP_016859595.1 | 4 | 562 | |||
XM_017004107.1 | XP_016859596.1 | 5 | 478 | |||
GenBank mRNA | CR749330.1 | - | 4 | 279 | ||
DC343949.1 | - | 4 | 354 |
Gene Symbol: RGPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144013.1 | NP_001137485.1 | 4 | 347 | ||
XM_017004738.1 | XP_016860227.1 | 5 | 396 | |||
XM_017004739.1 | XP_016860228.1 | 5 | 396 | |||
XM_017004740.1 | XP_016860229.1 | 4 | 321 |
Gene Symbol: RGPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537861.1 | - | 3 | 243 |
Gene Symbol: RGPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005054.2 | NP_005045.2 | 4 | 367 | ||
NM_032260.2 | NP_115636.1 | 5 | 823 | |||
XM_006712794.3 | XP_006712857.2 | 4 | 398 | |||
XM_017005080.1 | XP_016860569.1 | 4 | 387 | |||
XM_017005081.1 | XP_016860570.1 | 4 | 398 | |||
XM_017005082.1 | XP_016860571.1 | 3 | 215 | |||
GenBank mRNA | AL136868.1 | - | 5 | 824 | ||
EU794682.1 | - | 4 | 367 | |||
U64675.2 | - | 4 | 367 |
Gene Symbol: RGPD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037866.1 | NP_001032955.1 | 5 | 823 | ||
NM_001123363.3 | NP_001116835.1 | 5 | 823 | |||
XM_011511766.2 | XP_011510068.1 | 4 | 407 | |||
XM_011511767.2 | XP_011510069.1 | 4 | 407 | |||
XM_011511769.2 | XP_011510071.1 | 4 | 407 | |||
XM_011511770.2 | XP_011510072.1 | 4 | 407 | |||
XM_017004839.1 | XP_016860328.1 | 4 | 407 | |||
XM_017004840.1 | XP_016860329.1 | 4 | 480 | |||
XM_017004841.1 | XP_016860330.1 | 4 | 610 | |||
XM_017004842.1 | XP_016860331.1 | 4 | 387 | |||
XM_017004843.1 | XP_016860332.1 | 4 | 407 | |||
XM_017004844.1 | XP_016860333.1 | 3 | 215 |
Gene Symbol: RGPD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164463.1 | NP_001157935.1 | 4 | 482 | ||
XM_005263747.4 | XP_005263804.1 | 4 | 480 | |||
XM_011511733.2 | XP_011510035.1 | 4 | 480 | |||
XM_017004825.1 | XP_016860314.1 | 4 | 480 | |||
XM_017004826.1 | XP_016860315.1 | 4 | 480 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011240, AK041932, AK040322, AK145306.
Gene Ontology Categories:
