siRNA Details
Gene Symbol: FAM27C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027421.1 | 2 | 328 | |||
GenBank mRNA | BC002886.1 | - | 2 | 259 |
Gene Symbol: FAM27B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027422.1 | 2 | 328 | |||
GenBank mRNA | AK131029.1 | - | 2 | 211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
