siRNA Details
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 7 | 1557 | ||
NR_003494.1 | 6 | 1291 | ||||
XM_006716256.3 | XP_006716319.1 | 8 | 1652 | |||
XM_006716257.3 | XP_006716320.1 | 7 | 1722 | |||
XM_006716260.3 | XP_006716323.1 | 6 | 1466 | |||
XM_006716262.1 | XP_006716325.1 | 6 | 1309 | |||
XM_011543842.2 | XP_011542144.1 | 6 | 1731 | |||
XM_011543843.2 | XP_011542145.1 | 7 | 1670 | |||
XM_017013916.1 | XP_016869405.1 | 7 | 1698 | |||
XM_017013917.1 | XP_016869406.1 | 6 | 1619 | |||
XM_017013918.1 | XP_016869407.1 | 6 | 1591 | |||
XM_017013919.1 | XP_016869408.1 | 5 | 1180 | |||
GenBank mRNA | AK098138.1 | - | 7 | 1154 | ||
BC062615.1 | - | 7 | 1231 | |||
BC085016.1 | - | 8 | 1260 | |||
BC093808.1 | - | 1 | 106 | |||
BC093810.1 | - | 1 | 106 | |||
BC098427.1 | - | 7 | 1237 |
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273603.4 | XP_005273660.1 | 7 | 1587 | ||
XM_005273604.4 | XP_005273661.1 | 6 | 1506 | |||
XM_005273605.4 | XP_005273662.1 | 6 | 1407 | |||
XM_005273607.2 | XP_005273664.1 | 6 | 1445 | |||
XM_006716387.3 | XP_006716450.1 | 7 | 1608 | |||
XM_011544618.2 | XP_011542920.1 | 8 | 1659 | |||
XM_011544619.2 | XP_011542921.1 | 7 | 1626 | |||
XM_011544622.2 | XP_011542924.1 | 7 | 1528 | |||
XM_017013740.1 | XP_016869229.1 | 6 | 1589 | |||
XM_017013741.1 | XP_016869230.1 | 5 | 1526 | |||
XM_017013742.1 | XP_016869231.1 | 5 | 1508 |
Gene Symbol: FAM86C2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024249.1 | 4 | 1003 | |||
GenBank mRNA | AK055771.1 | - | 2 | 1656 |
Gene Symbol: FAM86DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024241.1 | 8 | 1649 | |||
GenBank mRNA | BC043342.2 | - | 7 | 1373 |
Gene Symbol: FAM86EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130740.1 | 4 | 1812 | |||
NR_130741.1 | 4 | 866 | ||||
NR_130742.1 | 4 | 816 | ||||
GenBank mRNA | BM718734.1 | - | 2 | 643 |
Gene Symbol: FAM86FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024254.1 | 5 | 1151 |
Gene Symbol: FAM86HP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024252.1 | 5 | 1262 | |||
GenBank mRNA | AK124448.1 | - | 3 | 1030 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
