siRNA Details
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005273603.4 | XP_005273660.1 | 7 | 1634 | ||
XM_005273604.4 | XP_005273661.1 | 6 | 1553 | |||
XM_005273605.4 | XP_005273662.1 | 6 | 1454 | |||
XM_005273607.2 | XP_005273664.1 | 6 | 1492 | |||
XM_006716387.3 | XP_006716450.1 | 7 | 1655 | |||
XM_011544618.2 | XP_011542920.1 | 8 | 1706 | |||
XM_011544619.2 | XP_011542921.1 | 7 | 1673 | |||
XM_011544622.2 | XP_011542924.1 | 7 | 1575 | |||
XM_017013740.1 | XP_016869229.1 | 6 | 1636 | |||
XM_017013741.1 | XP_016869230.1 | 5 | 1573 | |||
XM_017013742.1 | XP_016869231.1 | 5 | 1555 |
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 7 | 1604 | ||
NR_003494.1 | 6 | 1338 | ||||
XM_006716256.3 | XP_006716319.1 | 8 | 1699 | |||
XM_006716257.3 | XP_006716320.1 | 7 | 1769 | |||
XM_006716260.3 | XP_006716323.1 | 6 | 1513 | |||
XM_006716262.1 | XP_006716325.1 | 6 | 1356 | |||
XM_011543842.2 | XP_011542144.1 | 6 | 1778 | |||
XM_011543843.2 | XP_011542145.1 | 7 | 1717 | |||
XM_017013916.1 | XP_016869405.1 | 7 | 1745 | |||
XM_017013917.1 | XP_016869406.1 | 6 | 1666 | |||
XM_017013918.1 | XP_016869407.1 | 6 | 1638 | |||
XM_017013919.1 | XP_016869408.1 | 5 | 1227 | |||
GenBank mRNA | AK098138.1 | - | 7 | 1201 | ||
BC062615.1 | - | 7 | 1278 | |||
BC085016.1 | - | 8 | 1307 | |||
BC093808.1 | - | 1 | 153 | |||
BC093810.1 | - | 1 | 153 | |||
BC098427.1 | - | 7 | 1284 |
Gene Symbol: FAM86JP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024250.1 | 5 | 1117 | |||
NR_024251.1 | 5 | 1215 | ||||
GenBank mRNA | BC032772.1 | - | 5 | 1103 | ||
BC041080.1 | - | 5 | 1186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
