siRNA Details
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 5 | 490 | |||
NR_027776.2 | 4 | 387 | ||||
NR_027777.2 | 3 | 300 | ||||
GenBank mRNA | AK309660.1 | - | 4 | 335 | ||
BC052588.1 | - | 3 | 303 | |||
BF984173.1 | - | 4 | 386 | |||
D38436.1 | - | 5 | 471 | |||
D38439.1 | - | 3 | 299 | |||
D38501.1 | - | 3 | 299 | |||
D38502.1 | - | 5 | 471 |
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023383.1 | 9 | 1403 |
Gene Symbol: LOC105375355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012886.1 | XP_016868375.1 | 5 | 990 |
Gene Symbol: LOC441259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC075816.1 | - | 5 | 445 |
Gene Symbol: PMS2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003613.1 | 5 | 881 | |||
GenBank mRNA | AB017004.1 | - | 5 | 881 | ||
BC067809.1 | - | 5 | 513 | |||
BC110395.1 | - | 5 | 489 | |||
D38435.1 | - | 5 | 484 | |||
D38499.1 | - | 5 | 484 | |||
DA706474.1 | - | 1 | 15 |
Gene Symbol: PMS2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047753.1 | - | 4 | 340 |
Gene Symbol: PMS2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003614.3 | 4 | 364 |
Gene Symbol: PMS2P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_022007.2 | 4 | 375 | |||
NR_046297.1 | 4 | 375 | ||||
NR_046299.1 | 4 | 375 | ||||
GenBank mRNA | BC029368.1 | - | 4 | 352 | ||
BC029419.1 | - | 4 | 340 | |||
CB141754.1 | - | 4 | 340 | |||
D38438.1 | - | 4 | 346 | |||
D38500.1 | - | 4 | 346 | |||
DA885243.1 | - | 4 | 376 |
Gene Symbol: PMS2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130940.1 | 5 | 518 | |||
GenBank mRNA | AB017006.1 | - | 5 | 611 |
Gene Symbol: PMS2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028058.1 | 3 | 778 | |||
GenBank mRNA | BM845848.1 | - | 4 | 319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
