siRNA Details
Gene Symbol: RPL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000982.3 | NP_000973.2 | 1 | 433 | ||
GenBank mRNA | AF086172.1 | - | 1 | 332 | ||
AK130968.1 | - | 1 | 432 | |||
AK311795.1 | - | 1 | 430 | |||
BC001603.1 | - | 1 | 415 | 3543702 | ||
BC007505.2 | - | 1 | 396 | 2964441 | ||
BC019002.2 | - | 1 | 388 | |||
BC062981.1 | - | 1 | 413 | 6084268 | ||
BC070184.1 | - | 1 | 417 | 4281405 | ||
BC070323.1 | - | 1 | 430 | 4248655 | ||
BC070330.1 | - | 1 | 430 | 3931616 | ||
BC071902.1 | - | 1 | 413 | 4901492 | ||
BC104667.1 | - | 1 | 434 | 6724053 | ||
BX647669.1 | - | 1 | 432 | |||
CR457032.1 | - | 1 | 390 | |||
U14967.1 | - | 1 | 419 | |||
X89401.1 | - | 1 | 403 |
Gene Symbol: RPL21P19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209100.1 | - | 1 | 492 |
Gene Symbol: RPL21P28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026911.1 | 1 | 435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
