siRNA Details
Gene Symbol: GFPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244710.1 | NP_001231639.1 | 20 | 8559 | ||
NM_002056.3 | NP_002047.2 | 19 | 8505 | |||
GenBank mRNA | AF131846.1 | - | 1 | 1545 | ||
AF445027.1 | - | 1 | 3499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
