siRNA Details
Gene Symbol: TPM3P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003148.3 | 2 | 461 | |||
GenBank mRNA | AK094777.1 | - | 4 | 401 | ||
BC061910.1 | - | 1 | 272 | |||
BX647962.1 | - | 2 | 448 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
