siRNA Details
Gene Symbol: TPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043351.1 | NP_001036816.1 | 2, 3 | 402 | ||
NM_001043352.1 | NP_001036817.1 | 2, 3 | 402 | |||
NM_001043353.1 | NP_001036818.1 | 2, 3 | 402 | |||
NM_001278189.1 | NP_001265118.1 | 2, 3 | 402 | |||
NM_001278190.1 | NP_001265119.1 | 2, 3 | 402 | |||
NM_001278191.1 | NP_001265120.1 | 2, 3 | 248 | |||
NM_152263.3 | NP_689476.2 | 3, 4 | 500 | |||
NM_153649.3 | NP_705935.1 | 2, 3 | 402 | |||
NR_103461.1 | 2, 3 | 402 | ||||
GenBank mRNA | AF474157.1 | - | 2, 3 | 272 | ||
AK026559.1 | - | 2, 3 | 342 | |||
AK056921.1 | - | 2, 3 | 259 | |||
AK056997.1 | - | 3, 4 | 290 | |||
AK092690.1 | - | 3, 4 | 340 | |||
AK301674.1 | - | 2, 3 | 224 | |||
AK315504.1 | - | 2, 3 | 361 | |||
AL537561.3 | - | 2, 3 | 312 | |||
AY004867.1 | - | 2, 3 | 283 | |||
BC000771.2 | - | 2, 3 | 314 | 3506357 | ||
BC008407.1 | - | 3, 4 | 466 | 4245271 | ||
BC008425.1 | - | 3, 4 | 469 | 4246114 | ||
BC015403.1 | - | 2, 3 | 317 | 4389911 | ||
BC062740.1 | - | 3, 4 | 332 | 4271142 | ||
BC072428.1 | - | 2, 3 | 329 | 6472532 | ||
BF967161.1 | - | 2, 3 | 215 | |||
BG706843.1 | - | 2, 3 | 345 | |||
BQ933569.1 | - | 2, 3 | 369 | |||
BU956481.1 | - | 2, 3 | 328 | |||
BX419658.2 | - | 2, 3 | 315 | |||
CR997359.1 | - | 2, 3 | 293 | |||
DA209890.1 | - | 2, 3 | 251 | |||
DA472065.1 | - | 2, 3 | 402 | |||
DA894064.1 | - | 3, 4 | 290 | |||
EU668324.1 | - | 3, 4 | 340 | |||
KU178368.1 | - | 2, 3 | 272 | |||
KU178369.1 | - | 2, 3 | 272 | |||
KU178370.1 | - | 2, 3 | 272 | |||
X04201.1 | - | 3, 4 | 441 | |||
X04588.1 | - | 2, 3 | 324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
