siRNA Details
Gene Symbol: TPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043352.1 | NP_001036817.1 | 5, 6 | 663 | ||
NM_001278188.1 | NP_001265117.1 | 4, 5 | 529 | |||
NM_001278189.1 | NP_001265118.1 | 5, 6 | 663 | |||
NM_153649.3 | NP_705935.1 | 5, 6 | 663 | |||
NR_103461.1 | 5, 6 | 663 | ||||
GenBank mRNA | AB062125.1 | - | 1, 2 | 76 | ||
AF474157.1 | - | 5, 6 | 533 | |||
AK026559.1 | - | 5, 6 | 603 | |||
AK056889.1 | - | 1, 2 | 652 | |||
AK056921.1 | - | 5, 6 | 520 | |||
AK092712.1 | - | 4, 5 | 408 | |||
AK298678.1 | - | 4, 5 | 507 | |||
AK301674.1 | - | 5, 6 | 485 | |||
AK308007.1 | - | 4, 5 | 438 | |||
AK315504.1 | - | 5, 6 | 622 | |||
AY004867.1 | - | 5, 6 | 544 | |||
BC000771.2 | - | 5, 6 | 575 | 3506357 | ||
BC015403.1 | - | 5, 6 | 578 | 4389911 | ||
BC072428.1 | - | 5, 6 | 590 | 6472532 | ||
BU956481.1 | - | 5, 6 | 589 | |||
BX648485.1 | - | 5, 6 | 575 | |||
CR997359.1 | - | 5, 6 | 554 | |||
DA472065.1 | - | 5, 6 | 663 | |||
FJ224335.1 | - | 5, 6 | 575 | |||
KU178368.1 | - | 5, 6 | 533 | |||
M12128.1 | - | 1 | 161 | |||
X04588.1 | - | 5, 6 | 585 |
Gene Symbol: TPM3P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003148.3 | 2 | 653 | |||
GenBank mRNA | AK094777.1 | - | 4 | 593 | ||
BC061910.1 | - | 1 | 464 | |||
BX647962.1 | - | 2 | 640 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
