siRNA Details
Gene Symbol: OLFML2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282715.1 | NP_001269644.1 | 2 | 241 | ||
NM_182487.3 | NP_872293.2 | 5 | 822 | |||
XM_005251760.4 | XP_005251817.1 | 5 | 822 | |||
XM_006716989.2 | XP_006717052.1 | 4 | 714 | |||
GenBank mRNA | AB119055.1 | - | 5 | 709 | ||
AK074164.1 | - | 3 | 373 | |||
AK074709.1 | - | 4 | 714 | |||
AY358185.1 | - | 5 | 640 | |||
BC054001.1 | - | 2 | 241 | 5554812 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
