siRNA Details
Gene Symbol: SPDYE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166339.1 | NP_001159811.1 | 7 | 1288 | ||
XM_005250093.3 | XP_005250150.1 | 6 | 1856 | |||
XM_011515702.2 | XP_011514004.1 | 6 | 1856 |
Gene Symbol: SPDYE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031618.3 | NP_001026789.2 | 7 | 1288 | ||
XM_011516237.2 | XP_011514539.1 | 7 | 1287 | |||
XM_017012223.1 | XP_016867712.1 | 7 | 1287 | |||
GenBank mRNA | AK302217.1 | - | 7 | 1288 | ||
BC100973.1 | - | 4 | 526 | 40005226 | ||
DR731150.1 | - | 4 | 432 | |||
JQ023160.1 | - | 6 | 814 |
Gene Symbol: SPDYE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146210.2 | NP_001139682.2 | 6 | 1235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
