siRNA Details
Gene Symbol: TMPRSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135099.1 | NP_001128571.1 | 3, 4 | 404 | ||
NM_005656.3 | NP_005647.3 | 3, 4 | 366 | |||
XM_005261043.2 | XP_005261100.1 | 3, 4 | 366 | |||
XM_011529731.2 | XP_011528033.1 | 3, 4 | 461 | |||
XM_011529732.2 | XP_011528034.1 | 2, 3 | 288 | |||
GenBank mRNA | AF123453.1 | - | 1, 2 | 232 | ||
AF270487.1 | - | 3, 4 | 345 | |||
AF329454.1 | - | 3, 4 | 359 | |||
AK222784.1 | - | 2, 3 | 277 | |||
AK296860.1 | - | 3, 4 | 404 | |||
AK313338.1 | - | 4, 5 | 666 | |||
BC035623.1 | - | 3, 4 | 348 | |||
BC051839.1 | - | 3, 4 | 337 | 6199625 | ||
BP276431.1 | - | 3, 4 | 366 | |||
U75329.1 | - | 2, 3 | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
