siRNA Details
Gene Symbol: TMPRSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135099.1 | NP_001128571.1 | 10 | 1121 | ||
NM_005656.3 | NP_005647.3 | 10 | 1083 | |||
XM_005261043.2 | XP_005261100.1 | 9 | 963 | |||
XM_011529731.2 | XP_011528033.1 | 10 | 1178 | |||
XM_011529732.2 | XP_011528034.1 | 9 | 1005 | |||
XM_011529733.1 | XP_011528035.1 | 8 | 782 | |||
GenBank mRNA | AF123453.1 | - | 8 | 949 | ||
AF270487.1 | - | 10 | 1062 | |||
AF329454.1 | - | 10 | 1076 | |||
AK222784.1 | - | 9 | 994 | |||
AK291813.1 | - | 10 | 1038 | |||
AK296860.1 | - | 10 | 1121 | |||
AK313338.1 | - | 11 | 1383 | |||
BC035623.1 | - | 10 | 1060 | |||
BC051839.1 | - | 10 | 1054 | 6199625 | ||
U75329.1 | - | 9 | 1005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
