siRNA Details
Gene Symbol: TRIM49C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195234.1 | NP_001182163.1 | 3 | 621 | ||
XM_017018126.1 | XP_016873615.1 | 1 | 296 | |||
GenBank mRNA | BC126470.1 | - | 2 | 415 | 8992186 | |
JF968462.1 | - | 1 | 52 |
Gene Symbol: TRIM49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020358.2 | NP_065091.1 | 3 | 621 | ||
XM_017018027.1 | XP_016873516.1 | 1 | 296 | |||
GenBank mRNA | AB037682.1 | - | 3 | 621 | ||
BC075019.2 | - | 2 | 320 | 30915409 | ||
BC075020.2 | - | 2 | 320 | 30915620 | ||
BC126472.1 | - | 2 | 415 | 8992188 | ||
JF968461.1 | - | 1 | 52 |
Gene Symbol: TRIM49B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206626.1 | NP_001193555.1 | 1 | 292 | ||
XM_017017561.1 | XP_016873050.1 | 1 | 296 | |||
GenBank mRNA | JF968463.1 | - | 1 | 178 |
Gene Symbol: TRIM49D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206627.1 | NP_001193556.1 | 1 | 292 | ||
XM_011542816.2 | XP_011541118.1 | 2 | 408 |
Gene Symbol: TRIM49D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105522.1 | NP_001098992.1 | 2 | 406 |
Gene Symbol: TRIM51JP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JF968455.1 | - | 1 | 214 |
Gene Symbol: TRIM53AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028346.1 | 2 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
