siRNA Details
Gene Symbol: KRT16P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029393.1 | 1 | 169 | |||
GenBank mRNA | BC110641.1 | - | 1 | 152 | ||
DB013784.1 | - | 1 | 201 |
Gene Symbol: KRT16P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073414.1 | 1 | 171 |
Gene Symbol: KRT16P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029392.1 | 1 | 174 | |||
GenBank mRNA | AK304304.1 | - | 1 | 174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
