siRNA Details
Gene Symbol: OR6A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003696.2 | NP_003687.2 | 1 | 473 | ||
GenBank mRNA | BC101489.1 | - | 1 | 348 | 8068995 | |
BC101491.1 | - | 1 | 348 | 8068997 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
