siRNA Details
Gene Symbol: LOC102724197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724921.3 | XP_006724984.1 | 3 | 986 | ||
XM_006724924.3 | XP_006724987.1 | 3 | 986 | |||
XM_011546256.2 | XP_011544558.1 | 3 | 989 | |||
XM_017030133.1 | XP_016885622.1 | 4 | 551 | |||
XM_017030134.1 | XP_016885623.1 | 4 | 554 | |||
XM_017030135.1 | XP_016885624.1 | 4 | 979 | |||
XM_017030136.1 | XP_016885625.1 | 5 | 1021 | |||
XM_017030137.1 | XP_016885626.1 | 5 | 1018 | |||
XM_017030138.1 | XP_016885627.1 | 5 | 541 | |||
XM_017030139.1 | XP_016885628.1 | 5 | 544 | |||
XM_017030140.1 | XP_016885629.1 | 6 | 1011 | |||
XM_017030141.1 | XP_016885630.1 | 6 | 1008 | |||
XM_017030142.1 | XP_016885631.1 | 4 | 852 | |||
XM_017030143.1 | XP_016885632.1 | 3 | 987 | |||
XM_017030145.1 | XP_016885634.1 | 3 | 986 | |||
XM_017030146.1 | XP_016885635.1 | 3 | 1003 |
Gene Symbol: GGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724392.3 | XP_006724455.1 | 4 | 701 | ||
XM_017029179.1 | XP_016884668.1 | 5 | 1382 | |||
XM_017029180.1 | XP_016884669.1 | 4 | 840 | |||
XM_017029181.1 | XP_016884670.1 | 4 | 1278 | |||
XM_017029182.1 | XP_016884671.1 | 5 | 1382 | |||
XM_017029185.1 | XP_016884674.1 | 5 | 862 | |||
XM_017029186.1 | XP_016884675.1 | 7 | 1541 |
Gene Symbol: GGT3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003267.1 | 2 | 347 | |||
GenBank mRNA | BC143607.1 | - | 2 | 213 |
Gene Symbol: LOC102724823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030162.1 | XP_016885651.1 | 4 | 1020 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
