siRNA Details
Gene Symbol: GGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724392.3 | XP_006724455.1 | 4 | 614 | ||
XM_017029179.1 | XP_016884668.1 | 5 | 1295 | |||
XM_017029180.1 | XP_016884669.1 | 4 | 753 | |||
XM_017029181.1 | XP_016884670.1 | 4 | 1191 | |||
XM_017029182.1 | XP_016884671.1 | 5 | 1295 | |||
XM_017029185.1 | XP_016884674.1 | 5 | 775 | |||
XM_017029186.1 | XP_016884675.1 | 7 | 1454 |
Gene Symbol: GGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288833.1 | NP_001275762.1 | 5 | 863 | ||
NM_013421.2 | NP_038265.2 | 6 | 791 | |||
NM_013430.2 | NP_038347.2 | 5 | 551 | |||
GenBank mRNA | AL832738.1 | - | 767 | |||
BC025927.1 | - | 3 | 431 | 4548861 | ||
BU157907.1 | - | 5 | 534 | |||
BU164348.1 | - | 5 | 558 | |||
CR456494.1 | - | 2 | 141 | |||
J04131.1 | - | 6 | 733 | |||
J05235.1 | - | 4 | 808 | |||
M24087.1 | - | 1 | 66 | |||
M24903.1 | - | 5 | 551 | |||
X60069.1 | - | 3 | 422 |
Gene Symbol: GGT3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003267.1 | 2 | 260 | |||
GenBank mRNA | BC143607.1 | - | 2 | 126 |
Gene Symbol: GGT8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003503.1 | 1 | 666 | |||
GenBank mRNA | AK055838.1 | - | 1 | 666 |
Gene Symbol: LOC102724823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030162.1 | XP_016885651.1 | 4 | 933 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
