siRNA Details
Gene Symbol: PPP4R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318025.1 | NP_001304954.1 | 3 | 415 | ||
NM_001318026.1 | NP_001304955.1 | 4 | 357 | |||
NM_001318027.1 | NP_001304956.1 | 3 | 415 | |||
NM_174907.3 | NP_777567.1 | 4 | 586 | |||
GenBank mRNA | AF327345.1 | - | 4 | 403 | ||
AJ271448.2 | - | 1 | 984 | |||
AK289901.1 | - | 4 | 553 | |||
BC100281.1 | - | 4 | 546 | |||
BC110889.1 | - | 3 | 372 | 6503859 | ||
BC128136.1 | - | 4 | 362 | 40112443 | ||
BC128137.1 | - | 4 | 362 | 40112444 | ||
CB988831.1 | - | 3 | 424 | |||
HY034679.1 | - | 4 | 358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
