siRNA Details
Gene Symbol: CARMIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013838.1 | NP_001013860.1 | 32 | 3782 | ||
NM_001317026.1 | NP_001303955.1 | 33 | 3674 | |||
XM_011522874.1 | XP_011521176.1 | 33 | 3812 | |||
XM_011522875.2 | XP_011521177.2 | 33 | 3812 | |||
XM_017022953.1 | XP_016878442.1 | 33 | 3812 | |||
GenBank mRNA | AB113647.1 | - | 32 | 3782 | ||
BC043154.1 | - | 4 | 740 | |||
FJ026014.1 | - | 33 | 3554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
