siRNA Details
Gene Symbol: ZACN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180990.3 | NP_851321.2 | 7 | 815 | ||
GenBank mRNA | AB223030.1 | - | 7 | 729 | ||
AF512521.1 | - | 7 | 769 | |||
AK122638.1 | - | 7 | 1509 | |||
BC110597.1 | - | 7 | 769 | 40023955 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
