siRNA Details
Gene Symbol: PGAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002629.3 | NP_002620.1 | 1 | 203 | ||
GenBank mRNA | AK223096.1 | - | 1 | 49 | ||
AK292216.1 | - | 1 | 104 | |||
AK293023.1 | - | 1 | 48 | |||
AK295991.1 | - | 1 | 55 | |||
AK297491.1 | - | 1 | 73 | |||
AK312351.1 | - | 1 | 32 | |||
AY007118.1 | - | 1 | 40 | |||
BC000455.1 | - | 1 | 66 | 2821567 | ||
BC010038.1 | - | 1 | 58 | 3604026 | ||
BC011678.2 | - | 1 | 22 | 4299008 | ||
BC053356.1 | - | 1 | 39 | 5787678 | ||
BC062302.1 | - | 1 | 38 | 6584466 | ||
BC066959.1 | - | 1 | 71 | 5262935 | ||
BC073742.1 | - | 1 | 37 | 6297173 | ||
BC157873.1 | - | 1 | 55 | 8862699 | ||
CR541659.1 | - | 1 | 20 | |||
CR541744.1 | - | 1 | 20 | |||
DA270129.1 | - | 1 | 203 | |||
EU794644.1 | - | 1 | 32 | |||
J04173.1 | - | 1 | 51 |
Gene Symbol: PGAM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029891.2 | NP_001025062.1 | 1 | 20 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
