siRNA Details
Gene Symbol: PROS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000313.3 | NP_000304.2 | 2 | 434 | ||
NM_001314077.1 | NP_001301006.1 | 3 | 521 | |||
GenBank mRNA | AK292994.1 | - | 2 | 240 | ||
BC015801.1 | - | 2 | 410 | 3909023 | ||
BG702968.1 | - | 3 | 304 | |||
DA408604.1 | - | 2 | 427 | |||
DB076220.1 | - | 2 | 52 | |||
KU178283.1 | - | 2 | 93 | |||
KU178284.1 | - | 3 | 189 | |||
M14338.1 | - | 2 | 261 | |||
M15036.1 | - | 2 | 239 | |||
X12892.1 | - | 2 | 261 | |||
Y00692.1 | - | 2 | 216 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
