siRNA Details
Gene Symbol: KRTAP12-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198697.2 | NP_941970.2 | 1 | 287 | ||
GenBank mRNA | AB076361.1 | - | 1 | 287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
