siRNA Details
Gene Symbol: KRTAP12-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181686.1 | NP_859014.1 | 1 | 305 | ||
GenBank mRNA | AB076363.1 | - | 1 | 285 | ||
AJ566388.1 | - | 1 | 305 | |||
BC119713.2 | - | 1 | 294 | 40117204 | ||
BC120942.1 | - | 1 | 294 | 40115088 | ||
BC127648.1 | - | 1 | 289 | 40117199 |
Gene Symbol: KRTAP12-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198697.2 | NP_941970.2 | 1 | 313 | ||
GenBank mRNA | AB076361.1 | - | 1 | 313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
