siRNA Details
Gene Symbol: TUBA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006082.2 | NP_006073.2 | 4 | 727 | ||
GenBank mRNA | AF081484.1 | - | 3 | 506 | ||
AK026516.1 | - | 4 | 601 | |||
AK026632.1 | - | 4 | 601 | |||
AK056693.1 | - | 3 | 1509 | |||
AK057773.1 | - | 4 | 601 | |||
AK094717.1 | - | 3 | 2174 | |||
AK098357.1 | - | 4 | 601 | |||
AK098635.1 | - | 4 | 601 | |||
AK098771.1 | - | 4 | 601 | |||
AK289354.1 | - | 4 | 601 | |||
AK293337.1 | - | 2 | 226 | |||
BC000696.2 | - | 4 | 592 | 3349131 | ||
BC001128.1 | - | 4 | 585 | 2964447 | ||
BC006379.2 | - | 4 | 587 | 4100566 | ||
BC006481.1 | - | 4 | 587 | 2905305 | ||
BC008659.2 | - | 4 | 564 | 3871729 | ||
BC009314.2 | - | 4 | 586 | 4111591 | ||
BC009509.2 | - | 4 | 588 | 3533174 | ||
BC009512.2 | - | 4 | 585 | 3537301 | ||
BC009513.2 | - | 4 | 589 | 3537827 | ||
BC010494.1 | - | 4 | 587 | 3445903 | ||
BC011572.1 | - | 4 | 587 | 4561240 | ||
BC015883.2 | - | 4 | 545 | 3909374 | ||
BC017004.1 | - | 4 | 557 | 4430457 | ||
BC018948.2 | - | 4 | 587 | |||
BC030820.1 | - | 4 | 598 | 4277263 | ||
BC071904.1 | - | 4 | 592 | 6089096 | ||
DQ400107.1 | - | 4 | 591 | |||
K00558.1 | - | 4 | 573 |
Gene Symbol: TUBA1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303114.1 | NP_001290043.1 | 4 | 744 | ||
NM_001303115.1 | NP_001290044.1 | 6 | 977 | |||
NM_001303116.1 | NP_001290045.1 | 5 | 859 | |||
NM_001303117.1 | NP_001290046.1 | 5 | 862 | |||
NM_032704.4 | NP_116093.1 | 4 | 689 | |||
GenBank mRNA | AK223024.1 | - | 4 | 606 | ||
AK293589.1 | - | 4 | 744 | |||
AL539200.3 | - | 5 | 855 | |||
BC005946.1 | - | 4 | 606 | 4134187 | ||
BC011790.1 | - | 4 | 587 | 4025017 | ||
BC019298.2 | - | 4 | 568 | 2905506 | ||
BC021088.1 | - | 4 | 567 | 3543481 | ||
BC051297.1 | - | 4 | 582 | 3832363 | ||
BC063036.1 | - | 4 | 628 | 4397636 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009448, NM_011653, NM_011654, BC098321, AK169610, and others.
Gene Ontology Categories:
