siRNA Details
Gene Symbol: TUBA1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303114.1 | NP_001290043.1 | 4 | 1111 | ||
NM_001303115.1 | NP_001290044.1 | 6 | 1344 | |||
NM_001303116.1 | NP_001290045.1 | 5 | 1226 | |||
NM_001303117.1 | NP_001290046.1 | 5 | 1229 | |||
NM_032704.4 | NP_116093.1 | 4 | 1056 | |||
GenBank mRNA | AK223024.1 | - | 4 | 973 | ||
AK293589.1 | - | 4 | 1111 | |||
BC004949.1 | - | 4 | 953 | 3617714 | ||
BC005946.1 | - | 4 | 973 | 4134187 | ||
BC011790.1 | - | 4 | 954 | 4025017 | ||
BC019298.2 | - | 4 | 935 | 2905506 | ||
BC021088.1 | - | 4 | 934 | 3543481 | ||
BC033064.1 | - | 4 | 559 | 3926854 | ||
BC051297.1 | - | 4 | 949 | 3832363 | ||
BC063036.1 | - | 4 | 995 | 4397636 |
Gene Symbol: TUBA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270399.1 | NP_001257328.1 | 4 | 1778 | ||
NM_001270400.1 | NP_001257329.1 | 4 | 1306 | |||
NM_006009.3 | NP_006000.2 | 4 | 1218 | |||
GenBank mRNA | AF141347.1 | - | 4 | 952 | ||
AF141348.1 | - | 1 | 505 | |||
AK057842.1 | - | 4 | 972 | |||
AK289483.1 | - | 4 | 1145 | |||
AK298838.1 | - | 4 | 828 | |||
BC006468.1 | - | 4 | 927 | 3528520 | ||
BC050637.2 | - | 4 | 957 | 6050536 |
Gene Symbol: TUBA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006082.2 | NP_006073.2 | 4 | 1094 | ||
GenBank mRNA | AF081484.1 | - | 3 | 873 | ||
AK026516.1 | - | 4 | 968 | |||
AK026632.1 | - | 4 | 968 | |||
AK056693.1 | - | 3 | 1876 | |||
AK057773.1 | - | 4 | 968 | |||
AK094717.1 | - | 3 | 2541 | |||
AK098357.1 | - | 4 | 968 | |||
AK098635.1 | - | 4 | 968 | |||
AK098771.1 | - | 4 | 968 | |||
AK289354.1 | - | 4 | 968 | |||
BC000696.2 | - | 4 | 959 | 3349131 | ||
BC001128.1 | - | 4 | 952 | 2964447 | ||
BC006379.2 | - | 4 | 954 | 4100566 | ||
BC006481.1 | - | 4 | 954 | 2905305 | ||
BC008659.2 | - | 4 | 931 | 3871729 | ||
BC009314.2 | - | 4 | 953 | 4111591 | ||
BC009509.2 | - | 4 | 955 | 3533174 | ||
BC009512.2 | - | 4 | 952 | 3537301 | ||
BC009513.2 | - | 4 | 956 | 3537827 | ||
BC010494.1 | - | 4 | 954 | 3445903 | ||
BC011572.1 | - | 4 | 954 | 4561240 | ||
BC015883.2 | - | 4 | 912 | 3909374 | ||
BC017004.1 | - | 4 | 924 | 4430457 | ||
BC018948.2 | - | 4 | 954 | |||
BC021564.1 | - | 4 | 606 | 4579604 | ||
BC030820.1 | - | 4 | 965 | 4277263 | ||
BC071904.1 | - | 4 | 959 | 6089096 | ||
DQ400107.1 | - | 4 | 958 | |||
Z36832.1 | - | 1 | 183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
