siRNA Details
Gene Symbol: TUBA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006082.2 | NP_006073.2 | 4 | 1684 | ||
GenBank mRNA | AK026516.1 | - | 4 | 1558 | ||
AK026632.1 | - | 4 | 1558 | |||
AK056693.1 | - | 3 | 2466 | |||
AK057773.1 | - | 4 | 1558 | |||
AK094717.1 | - | 3 | 3131 | |||
AK098357.1 | - | 4 | 1558 | |||
AK098635.1 | - | 4 | 1558 | |||
AK098771.1 | - | 4 | 1558 | |||
AK289354.1 | - | 4 | 1558 | |||
AK293337.1 | - | 2 | 1183 | |||
BC000696.2 | - | 4 | 1549 | 3349131 | ||
BC001128.1 | - | 4 | 1542 | 2964447 | ||
BC006379.2 | - | 4 | 1544 | 4100566 | ||
BC006481.1 | - | 4 | 1544 | 2905305 | ||
BC008659.2 | - | 4 | 1521 | 3871729 | ||
BC009314.2 | - | 4 | 1543 | 4111591 | ||
BC009509.2 | - | 4 | 1545 | 3533174 | ||
BC009512.2 | - | 4 | 1542 | 3537301 | ||
BC009513.2 | - | 4 | 1546 | 3537827 | ||
BC010494.1 | - | 4 | 1544 | 3445903 | ||
BC011572.1 | - | 4 | 1544 | 4561240 | ||
BC015883.2 | - | 4 | 1502 | 3909374 | ||
BC017004.1 | - | 4 | 1514 | 4430457 | ||
BC018948.2 | - | 4 | 1544 | |||
BC021564.1 | - | 4 | 1196 | 4579604 | ||
BC030820.1 | - | 4 | 1555 | 4277263 | ||
BC071904.1 | - | 4 | 1549 | 6089096 | ||
DQ400107.1 | - | 4 | 1548 | |||
K00558.1 | - | 4 | 1530 | |||
Z36832.1 | - | 1 | 770 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
