siRNA Details
Gene Symbol: MAPK8IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005456.3 | NP_005447.1 | 10 | 2240 | ||
XM_005253226.3 | XP_005253283.1 | 10 | 2285 | |||
GenBank mRNA | AB209720.1 | - | 6 | 732 | ||
AF007134.1 | - | 6 | 516 | |||
AF074091.1 | - | 10 | 1915 | |||
AK293637.1 | - | 9 | 1183 | |||
AK295942.1 | - | 7 | 1361 | |||
BC068470.1 | - | 10 | 2085 | 5260971 |
Gene Symbol: LOC644172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026901.3 | 1 | 773 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
