siRNA Details
Gene Symbol: LTB4R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF277230.1 | - | 1 | 29 |
Gene Symbol: NOP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286367.1 | NP_001273296.1 | 10 | 5733 | ||
NM_174913.2 | NP_777573.1 | 10 | 5899 | |||
GenBank mRNA | AB095941.1 | - | 10 | 5891 | ||
BC015481.1 | - | 1 | 1393 | |||
BX647311.1 | - | 10 | 5866 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
