siRNA Details
Gene Symbol: BRCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007294.3 | NP_009225.1 | 3 | 350 | ||
NM_007298.3 | NP_009229.2 | 2 | 137 | |||
NM_007299.3 | NP_009230.2 | 3 | 312 | |||
NM_007300.3 | NP_009231.2 | 3 | 350 | |||
NR_027676.1 | 3 | 279 | ||||
GenBank mRNA | AF005068.1 | - | 3 | 244 | ||
AK307553.1 | - | 3 | 231 | |||
AK308084.1 | - | 3 | 279 | |||
AL701927.1 | - | 3 | 350 | |||
AY354539.2 | - | 2 | 182 | |||
AY751490.1 | - | 2 | 118 | |||
BC030969.1 | - | 3 | 237 | |||
BC072418.1 | - | 3 | 312 | |||
BC085615.1 | - | 3 | 220 | |||
BC106745.1 | - | 3 | 167 | |||
BC114511.1 | - | 3 | 167 | |||
BC115037.1 | - | 3 | 255 | 7961446 | ||
JN686490.1 | - | 2 | 118 | |||
KP272105.1 | - | 38 | ||||
U14680.1 | - | 3 | 237 | |||
U64805.1 | - | 2 | 118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
